accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 546-88-3

CH3ONHHO

CAS RN 546-88-3

IUPAC Name: N-hydroxyacetamide
Synonyms: acetohydroxamic acid 醋羟胺酸 acido acetohidroxamico acetohydroxamate acide acetohydroxamique AHA acetohydroximic acid N-acetyl hydroxyacetamide cetohyroxamic acid acethydroxamsaeure
Molecular Weight (g/mol): 75.07
Molecular Formula: C2H5NO2
InChi Key: RRUDCFGSUDOHDG-UHFFFAOYSA-N
SMILES: CC(=O)NO
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results