accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 546122-71-8

IUPAC Name: magnesium(2+) (2,4-difluorophenyl)methanide bromide
Synonyms: magnesium(2+) (2,4-difluorophenyl)methanide bromide
Molecular Weight (g/mol): 231.32
Molecular Formula: C7H5BrF2Mg
InChi Key: KHKZBHQZRCTDOD-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].[CH2-]C1=C(F)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results