accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HNNBr

CAS RN 54624-57-6

IUPAC Name: 2-bromo-1H-1,3-benzodiazole
Synonyms: 2-bromo-1H-1,3-benzodiazole
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: PHPYXVIHDRDPDI-UHFFFAOYSA-N
SMILES: BrC1=NC2=CC=CC=C2N1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results