accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5466-88-6

IUPAC Name: 3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms: 2H-1,4-benzoxazin-3(4H)-one
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: QRCGFTXRXYMJOS-UHFFFAOYSA-N
SMILES: O=C1COC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results