accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHNOO(E)

CAS RN 54673-07-3

IUPAC Name: (2E)-2-cyano-3-(3-hydroxyphenyl)prop-2-enoate
Synonyms: (2E)-2-cyano-3-(3-hydroxyphenyl)prop-2-enoate
Molecular Weight (g/mol): 188.16
Molecular Formula: C10H6NO3
InChi Key: HPLNTJVXWMJLNJ-XBXARRHUSA-M
SMILES: OC1=CC=CC(\C=C(/C#N)C([O-])=O)=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results