accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHOHO(E)

CAS RN 5469-45-4

IUPAC Name: (2E)-2-acetamido-3-phenylprop-2-enoic acid
Synonyms: (2E)-2-acetamido-3-phenylprop-2-enoic acid
Molecular Weight (g/mol): 205.21
Molecular Formula: C11H11NO3
InChi Key: XODAOBAZOQSFDS-JXMROGBWSA-N
SMILES: CC(=O)N\C(=C\C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results