accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2ClNN

CAS RN 5469-69-2

IUPAC Name: 6-chloropyridazin-3-amine
Synonyms: 3-CL-6-pyridazinamine
Molecular Weight (g/mol): 129.55
Molecular Formula: C4H4ClN3
InChi Key: DTXVKPOKPFWSFF-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)N=N1
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (5)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)
  • (3)