accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOCH3N

CAS RN 5470-70-2

IUPAC Name: methyl 6-methylpyridine-3-carboxylate
Synonyms: methyl 6-methylpyridine-3-carboxylate
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: VYPPZXZHYDSBSJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C)N=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)