accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5471-51-2

IUPAC Name: 4-(4-hydroxyphenyl)butan-2-one
Synonyms: raspberry ketone
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: NJGBTKGETPDVIK-UHFFFAOYSA-N
SMILES: CC(=O)CCC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)