accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5471-63-6

IUPAC Name: 1,3-diphenyl-2-benzofuran
Synonyms: 1,3-diphenyl-2-benzofuran
Molecular Weight (g/mol): 270.33
Molecular Formula: C20H14O
InChi Key: ZKSVYBRJSMBDMV-UHFFFAOYSA-N
SMILES: O1C(=C2C=CC=CC2=C1C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (7)
  • (2)
Percent Purity 
  • (5)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (7)
  • (2)
  • (5)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)