accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 548-24-3

IUPAC Name: disodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Synonyms: eosin B
Molecular Weight (g/mol): 624.06
Molecular Formula: C20H6Br2N2Na2O9
InChi Key: GYYTYUGGVBYJHE-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(C=C12)[N+]([O-])=O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
Color 
  • (2)
  • (2)
Grade 
  • (4)
pH 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)