CAS RN 54827-17-7
3,3',5,5'-Tetramethylbenzidine, 98%
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 MDL Number: MFCD00007748 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine, 99+%
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, high sensitivity, Thermo Scientific Chemicals
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 MDL Number: MFCD00007748 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, standard sensitivity
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 MDL Number: MFCD00007748 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, standard sensitivity, Thermo Scientific Chemicals
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 MDL Number: MFCD00007748 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, high sensitivity, Thermo Scientific Chemicals
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 MDL Number: MFCD00007748 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine, MP Biomedicals™
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C
3,3',5,5'-Tetramethylbenzidine, TRC
CAS: 54827-17-7 Molecular Formula: C16 H20 N2 Molecular Weight (g/mol): 240.34 Synonym: [1,1'-Biphenyl]-4,4'-diamine, 3,3',5,5'-tetramethyl- (9CI, ACI),3,3',5,5'-Tetramethyl[1,1'-biphenyl]-4,4'-diamine (ACI),Benzidine, 3,3',5,5'-tetramethyl- (6CI),3,3',5,5'-Tetramehylbenzidine,3,3',5,5'-Tetramethyl-4,4'-diaminobiphenyl,3,3',5,5'-Tetramethylbenzidine,3,3',5,5'-Tetramethylbiphenyl-4,4'-diamine,3,5,3',5'-Tetramethylbenzidine,4,4'-Diamino-3,3',5,5'-tetramethyl-1,1'-biphenyl,BM blue,ColorBurst Blue,Enhanced K-Blue,K-Blue,K-Blue Max,Sure Blue TMB,TMB,TMB Blotting Plus,TMB substrate IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: Cc1cc(cc(C)c1N)c2cc(C)c(N)c(C)c2
3,3',5,5'-Tetramethylbenzidine aq. soln., Thermo Scientific™
CAS: 54827-17-7 Molecular Formula: C16H20N2 Molecular Weight (g/mol): 240.35 MDL Number: MFCD00007748 InChI Key: UAIUNKRWKOVEES-UHFFFAOYSA-N Synonym: 3,3',5,5'-tetramethylbenzidine,3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine,3,5,3',5'-tetramethylbenzidine,tmb,3,3',5,5'-tetramethyl benzidine,tmb substrate,bm blue,ccris 4727,tmb plus,tmb, insoluble PubChem CID: 41206 IUPAC Name: 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline SMILES: CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C