accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 54856-23-4

IUPAC Name: bis(methanesulfonic acid); methyl[2-(pyridin-2-yl)ethyl]amine
Synonyms: betahistine; bis(methanesulfonic acid)
Molecular Weight (g/mol): 328.4
Molecular Formula: C10H20N2O6S2
InChi Key: ZBJJDYGJCNTNTH-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O.CS(O)(=O)=O.CNCCC1=CC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results