accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 54897-62-0

IUPAC Name: 4-[ethyl(nitroso)amino]butan-1-ol
Synonyms: 4-[ethyl(nitroso)amino]butan-1-ol
Molecular Weight (g/mol): 146.19
Molecular Formula: C6H14N2O2
InChi Key: SAWPRHGGVOPPSR-UHFFFAOYSA-N
SMILES: CCN(CCCCO)N=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results