CAS RN 54914-53-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Liothyronine Methyl Ester, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
3,3’,5-Triiodo-L-thyronine Methyl Ester, TRC
CAS: 54914-53-3 Molecular Formula: C16 H14 I3 N O4 Molecular Weight (g/mol): 665 Synonym: O-(4-hydroxy-3-iodophenyl)-3,5-diiodo-L-Tyrosine methyl ester,3,5,3'-Triiodo-L-thyronine methyl ester,L-Triiodothyronine methyl ester IUPAC Name: methyl (2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate SMILES: COC(=O)[C@@H](N)Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1