CAS RN 54947-74-9
IUPAC Name:
(4R)-4-methyloctanoate
Synonyms:
(4R)-4-methyloctanoate
Molecular Weight (g/mol):
157.23
Molecular Formula:
C9H17O2
InChi Key:
LEGGANXCVQPIAI-MRVPVSSYSA-M
SMILES:
CCCC[C@@H](C)CCC([O-])=O
1
–
2
of
2
results
1
4-Methyloctanoic acid, 98%
CAS: 54947-74-9 Molecular Formula: C9H17O2 Molecular Weight (g/mol): 157.23 MDL Number: MFCD00051938 InChI Key: LEGGANXCVQPIAI-MRVPVSSYSA-M Synonym: octanoic acid, 4-methyl,4-methyl-octanoic acid,4-methylcaprylic acid,+/--4-methyloctanoic acid,4-methyl-n-octanoic acid,4-methyl octanoic acid,octanoic acid, 4-methyl-, r,fema no. 3575,acmc-20mssw,acmc-20mssx PubChem CID: 62089 IUPAC Name: 4-methyloctanoic acid SMILES: CCCC[C@@H](C)CCC([O-])=O
2
Hircinoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.