accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 54970-72-8

IUPAC Name: sodium 3,5-dichloro-2-hydroxybenzene-1-sulfonic acid
Synonyms: sodium 3,5-dichloro-2-hydroxybenzenesulfonic acid
Molecular Weight (g/mol): 266.04
Molecular Formula: C6H4Cl2NaO4S
InChi Key: NMWCVZCSJHJYFW-UHFFFAOYSA-N
SMILES: [Na+].OC1=C(Cl)C=C(Cl)C=C1S(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results