accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 55-18-5

CH3NH3CNO

CAS RN 55-18-5

IUPAC Name: diethyl(nitroso)amine
Synonyms: DANA NDEA DEN N-nitroso-diaethylamine ccris 239 diaethylnitrosamin rcra waste number U174 diethylnitrosamide N-nitroso-N,N-diethylamine den (mutagen)
Molecular Weight (g/mol): 102.14
Molecular Formula: C4H10N2O
InChi Key: WBNQDOYYEUMPFS-UHFFFAOYSA-N
SMILES: CCN(CC)N=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results