accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 55-22-1

IUPAC Name: pyridine-4-carboxylic acid
Synonyms: isonicotinic acid acide iso-nicotinique INA 4-picolinic acid gamma-picolinic acid
Molecular Weight (g/mol): 123.11
Molecular Formula: C6H5NO2
InChi Key: TWBYWOBDOCUKOW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (8)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results