accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 550-24-3

H3COHOOHO

CAS RN 550-24-3

IUPAC Name: 2,5-dihydroxy-3-undecylcyclohexa-2,5-diene-1,4-dione
Synonyms: embelin embelic acid emberine potassium embelate
Molecular Weight (g/mol): 294.39
Molecular Formula: C17H26O4
InChi Key: IRSFLDGTOHBADP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC1=C(O)C(=O)C=C(O)C1=O
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)