accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 550-44-7

IUPAC Name: 2-methyl-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-methylisoindole-1,3-dione
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: ZXLYYQUMYFHCLQ-UHFFFAOYSA-N
SMILES: CN1C(=O)C2=CC=CC=C2C1=O