accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 55028-72-3

IUPAC Name: sodium (5Z)-7-[(1R,3R,5S)-2-[(1E,3R)-4-(3-chlorophenoxy)-3-hydroxybut-1-en-1-yl]-3,5-dihydroxycyclopentyl]hept-5-enoate
Synonyms: sodium (5Z)-7-[(1R,3R,5S)-2-[(1E,3R)-4-(3-chlorophenoxy)-3-hydroxybut-1-en-1-yl]-3,5-dihydroxycyclopentyl]hept-5-enoate
Molecular Weight (g/mol): 446.9
Molecular Formula: C22H28ClNaO6
InChi Key: IFEJLMHZNQJGQU-UDEWSXLWSA-M
SMILES: [Na+].O[C@@H](COC1=CC=CC(Cl)=C1)\C=C\C1[C@H](O)C[C@H](O)[C@@H]1C\C=C/CCCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results