accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHBrO

CAS RN 55082-33-2

IUPAC Name: 2-benzoyl-4-bromophenol
Synonyms: 2-benzoyl-4-bromophenol
Molecular Weight (g/mol): 277.12
Molecular Formula: C13H9BrO2
InChi Key: IVIICRNXAGUXLR-UHFFFAOYSA-N
SMILES: OC1=CC=C(Br)C=C1C(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results