accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2FF

CAS RN 5509-65-9

IUPAC Name: 2,6-difluoroaniline
Synonyms: 2,6-difluoroaniline
Molecular Weight (g/mol): 129.11
Molecular Formula: C6H5F2N
InChi Key: ODUZJBKKYBQIBX-UHFFFAOYSA-N
SMILES: NC1=C(F)C=CC=C1F
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)