accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 552-16-9

IUPAC Name: 2-nitrobenzoic acid
Synonyms: 2-nitrobenzoic acid
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: SLAMLWHELXOEJZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results