accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 552-30-7

IUPAC Name: 1,3-dioxo-1,3-dihydro-2-benzofuran-5-carboxylic acid
Synonyms: trimellitic anhydride anhydrotrimellic acid anhydrotrimellitic acid trimellitic acid 1,2-anhydride trimellic acid 1,2-anhydride TMAN trimellic acid anhydride trimellitic acid anhydride
Molecular Weight (g/mol): 192.13
Molecular Formula: C9H4O5
InChi Key: SRPWOOOHEPICQU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C(=O)OC(=O)C2=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (4)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (3)