accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 552-46-5

HClNH2

CAS RN 552-46-5

IUPAC Name: hydrogen naphthalen-1-amine chloride
Synonyms: hydrogen α naphthylamine chloride
Molecular Weight (g/mol): 179.65
Molecular Formula: C10H10ClN
InChi Key: FOKKJVHTXPJHEN-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=C2C=CC=CC2=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)