accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 552-66-9

IUPAC Name: 3-(4-hydroxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4H-chromen-4-one
Synonyms: daidzin daidzein 7-O-β-D-glucoside
Molecular Weight (g/mol): 416.38
Molecular Formula: C21H20O9
InChi Key: KYQZWONCHDNPDP-QNDFHXLGSA-N
SMILES: OC[C@H]1O[C@@H](OC2=CC=C3C(=O)C(=COC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@@H](O)[C@@H]1O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)