accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 552-89-6

IUPAC Name: 2-nitrobenzaldehyde
Synonyms: 2-nitrobenzaldehyde O-nitrobenzaldehyde
Molecular Weight (g/mol): 151.12
Molecular Formula: C7H5NO3
InChi Key: CMWKITSNTDAEDT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1C=O
Molecular Weight (g/mol) 
  • (7)
  • (6)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (7)
  • (6)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)