CAS RN 5521-38-0
IUPAC Name:
2-[4-(4-nitrophenyl)piperazin-1-yl]ethan-1-ol
Synonyms:
2-[4-(4-nitrophenyl)piperazin-1-yl]ethanol
Molecular Weight (g/mol):
251.29
Molecular Formula:
C12H17N3O3
InChi Key:
WVNPWSRYQDRSIS-UHFFFAOYSA-N
SMILES:
OCCN1CCN(CC1)C1=CC=C(C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
2-[4-(4-Nitrophenyl)-1-piperazinyl]ethanol, 97%, Thermo Scientific™
CAS: 5521-38-0 Molecular Formula: C12H17N3O3 Molecular Weight (g/mol): 251.286 MDL Number: MFCD00511837 InChI Key: WVNPWSRYQDRSIS-UHFFFAOYSA-N Synonym: 2-4-4-nitrophenyl piperazin-1-yl ethanol,2-4-4-nitrophenyl piperazino-1-ethanol,2-4-4-nitrophenyl piperazin-1-yl ethan-1-ol,2-4-4-nitro-phenyl-piperazin-1-yl-ethanol,2-4-4-nitrophenyl piperazinyl ethan-1-ol,nitrophenylpiperazinoethanol,cambridge id 5139370,4-4-nitrophenyl-1-piperazine-ethanol,2-4-4-nitrophenyl-1-piperazinyl ethanol PubChem CID: 2763936 IUPAC Name: 2-[4-(4-nitrophenyl)piperazin-1-yl]ethanol SMILES: C1CN(CCN1CCO)C2=CC=C(C=C2)[N+](=O)[O-]