accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrOHN

CAS RN 552330-86-6

IUPAC Name: 5-bromo-2,3-dihydro-1H-isoindol-1-one
Synonyms: 5-bromo-2,3-dihydroisoindol-1-one
Molecular Weight (g/mol): 212.05
Molecular Formula: C8H6BrNO
InChi Key: WJNKJYJCWXMBNV-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)C(=O)NC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results