accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOH3CCH3CH3

CAS RN 5524-57-2

IUPAC Name: ethyl 2-oxo-2-(2,4,6-trimethylphenyl)acetate
Synonyms: ethyl 2-oxo-2-(2,4,6-trimethylphenyl)acetate
Molecular Weight (g/mol): 220.27
Molecular Formula: C13H16O3
InChi Key: XRSPPFOBPRPYLD-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)C1=C(C)C=C(C)C=C1C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results