accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 55289-36-6

IUPAC Name: 3-bromo-2-methylaniline
Synonyms: 3-bromo-2-methylaniline
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: IILVSKMKMOJHMA-UHFFFAOYSA-N
SMILES: CC1=C(N)C=CC=C1Br
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results