accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClCH3NH3CNCH3NH2N

CAS RN 553-24-2

IUPAC Name: hydrogen N2,N2,7-trimethylphenazine-2,8-diamine chloride
Synonyms: neutral red
Molecular Weight (g/mol): 288.78
Molecular Formula: C15H17ClN4
InChi Key: PGSADBUBUOPOJS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=NC2=C1
Quantity 
  • (1)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (11)
Color 
  • (5)
  • (6)
Grade 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (9)
  • (2)
  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (11)
  • (5)
  • (6)
  • (2)
  • (4)