accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 553-26-4

IUPAC Name: 4,4'-bipyridine
Synonyms: bipyridine 4,4-bipyridyl 4,4-bipyridin 4,4-dipyridyl
Molecular Weight (g/mol): 156.19
Molecular Formula: C10H8N2
InChi Key: MWVTWFVJZLCBMC-UHFFFAOYSA-N
SMILES: C1=CC(=CC=N1)C1=CC=NC=C1
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results