accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOHNN

CAS RN 55317-53-8

IUPAC Name: 2-(1H-pyrazol-1-yl)benzoic acid
Synonyms: 2-(pyrazol-1-yl)benzoic acid
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: MHACZVWKWUMHRR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1N1C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results