accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 554-68-7

IUPAC Name: hydrogen triethylamine chloride
Synonyms: hydrogen triethylamine chloride
Molecular Weight (g/mol): 137.65
Molecular Formula: C6H16ClN
InChi Key: ILWRPSCZWQJDMK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCN(CC)CC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)