accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 554-73-4

IUPAC Name: sodium 4-{2-[4-(phenylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
Synonyms: acid orange 5
Molecular Weight (g/mol): 375.38
Molecular Formula: C18H14N3NaO3S
InChi Key: MLVYOYVMOZFHIU-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(NC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results