accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 554-84-7

IUPAC Name: 3-nitrophenol
Synonyms: 3-nitrophenol
Molecular Weight (g/mol): 139.11
Molecular Formula: C6H5NO3
InChi Key: RTZZCYNQPHTPPL-UHFFFAOYSA-N
SMILES: OC1=CC=CC(=C1)[N+]([O-])=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (3)
Boiling Point 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (6)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (6)
  • (1)