accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOO

CAS RN 55453-87-7

IUPAC Name: 2-{2-oxo-9-oxatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-5-yl}acetic acid
Synonyms: artil isoxepacum isoxepac
Molecular Weight (g/mol): 268.27
Molecular Formula: C16H12O4
InChi Key: QFGMXJOBTNZHEL-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C2OCC3=CC=CC=C3C(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results