accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BCH3CH3HOOH

CAS RN 55499-44-0

IUPAC Name: (2,4-dimethylphenyl)boronic acid
Synonyms: 2,4-dimethylphenylboronic acid
Molecular Weight (g/mol): 149.98
Molecular Formula: C8H11BO2
InChi Key: TYONHSPZXLFWKI-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)