accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 555-16-8

IUPAC Name: 4-nitrobenzaldehyde
Synonyms: 4-nitrobenzaldehyde
Molecular Weight (g/mol): 151.12
Molecular Formula: C7H5NO3
InChi Key: BXRFQSNOROATLV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=O)C=C1
Quantity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results