accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 555-60-2

IUPAC Name: N-(3-chlorophenyl)-1-cyanomethanecarbohydrazonoyl cyanide
Synonyms: CCCP CCP C9H5ClN4 carbonyl cyanide meta chlorophenyl hydrazone carbonyl cyanide m chlorophenyl hydrazone
Molecular Weight (g/mol): 204.62
Molecular Formula: C9H5ClN4
InChi Key: UGTJLJZQQFGTJD-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(NN=C(C#N)C#N)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)