accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 55512-05-5

IUPAC Name: N-(3-formylphenyl)methanesulfonamide
Synonyms: N-(3-formylphenyl)methanesulfonamide
Molecular Weight (g/mol): 199.22
Molecular Formula: C8H9NO3S
InChi Key: CBDSSTWZEANOCR-UHFFFAOYSA-N
SMILES: CS(=O)(=O)NC1=CC=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results