accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 55579-72-1

OOF

CAS RN 55579-72-1

IUPAC Name: 5-(4-fluorophenyl)cyclohexane-1,3-dione
Synonyms: 5-(4-fluorophenyl)cyclohexane-1,3-dione
Molecular Weight (g/mol): 206.22
Molecular Formula: C12H11FO2
InChi Key: LBTLJACXBCUFEF-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1CC(=O)CC(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results