accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NClNH2ClN

CAS RN 55583-59-0

IUPAC Name: dichloropyrimidine-2,5-diamine
Synonyms: dichloropyrimidine-2,5-diamine
Molecular Weight (g/mol): 179
Molecular Formula: C4H4Cl2N4
InChi Key: ZXWGHENZKVQKPX-UHFFFAOYSA-N
SMILES: NC1=NC(Cl)=C(N)C(Cl)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results