accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

KCH3ONSOOO

CAS RN 55589-62-3

IUPAC Name: potassium 6-methyl-2,4-dioxo-4H-1,2λ⁶,3-oxathiazin-2-olate
Synonyms: potassium acesulfame
Molecular Weight (g/mol): 201.24
Molecular Formula: C4H4KNO4S
InChi Key: JLEKLYQXZHJOTQ-UHFFFAOYSA-M
SMILES: [K+].CC1=CC(=O)N=S([O-])(=O)O1
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results