accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClH2NONHOHO

CAS RN 556-50-3

IUPAC Name: hydrogen 2-(2-aminoacetamido)acetic acid chloride
Synonyms: dipeptide-15
Molecular Weight (g/mol): 168.58
Molecular Formula: C4H9ClN2O3
InChi Key: YHBAZQDEMYQPJL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC(=O)NCC(O)=O
Molecular Weight (g/mol) 
  • (15)
  • (5)
Quantity 
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (3)
Physical Form 
  • (6)
  • (1)
pH 
  • (1)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (9)
  • (1)
  • (3)
  • (15)
  • (5)
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)