accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOO

CAS RN 556-52-5

IUPAC Name: (oxiran-2-yl)methanol
Synonyms: (+-)-glycidol glycidol, (+-)-isomer (r)-glycidol glycidol, (R)-isomer glycidol, (R)- allyl alcohol oxide glycide glycidol, (r)- glycidol, (r)-isomer monoepoxide glycidol
Molecular Weight (g/mol): 74.08
Molecular Formula: C3H6O2
InChi Key: CTKINSOISVBQLD-UHFFFAOYNA-N
SMILES: OCC1CO1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)