accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClCH3H2N

CAS RN 556-53-6

IUPAC Name: hydrogen propan-1-amine chloride
Synonyms: hydrogen propylamine chloride
Molecular Weight (g/mol): 95.57
Molecular Formula: C3H10ClN
InChi Key: PYNUOAIJIQGACY-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCCN
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results